siRNA Details
| Gene Symbol: Andpro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012718.2 | NP_036850.1 | 4 | 559 | ||
| GenBank mRNA | BC065311.1 | - | 1 | 6077 | ||
| M27901.1 | - | 4 | 543 | |||
| M58167.1 | - | 4 | 560 | |||
| S48988.1 | - | 2 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Andpro| Gene Symbol: Andpro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012718.2 | NP_036850.1 | 4 | 559 | ||
| GenBank mRNA | BC065311.1 | - | 1 | 6077 | ||
| M27901.1 | - | 4 | 543 | |||
| M58167.1 | - | 4 | 560 | |||
| S48988.1 | - | 2 | 99 | |||