siRNA Details
| Gene Symbol: Cyb5r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138877.1 | NP_620232.1 | 5 | 447 | ||
| XM_006242065.2 | XP_006242127.1 | 5 | 475 | |||
| GenBank mRNA | BC062066.1 | - | 5 | 463 | 5599255 | |
| D00636.1 | - | 5 | 447 | |||
| J03867.1 | - | 5 | 442 | |||

Cyb5r3
Genomic Map