siRNA Details
| Gene Symbol: Eln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012722.1 | NP_036854.1 | 24, 25 | 1692 | ||
| GenBank mRNA | BC085910.1 | - | 24, 25 | 1692 | 7097661 | |
| M60647.1 | - | 24, 25 | 1672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eln| Gene Symbol: Eln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012722.1 | NP_036854.1 | 24, 25 | 1692 | ||
| GenBank mRNA | BC085910.1 | - | 24, 25 | 1692 | 7097661 | |
| M60647.1 | - | 24, 25 | 1672 | |||