siRNA Details
| Gene Symbol: Pigr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012723.4 | NP_036855.3 | 963 | |||
| GenBank mRNA | X15741.1 | - | 3 | 857 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pigr| Gene Symbol: Pigr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012723.4 | NP_036855.3 | 963 | |||
| GenBank mRNA | X15741.1 | - | 3 | 857 | ||