siRNA Details
| Gene Symbol: Lipa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012732.3 | NP_036864.3 | 10 | 1448 | ||
| XM_006231275.2 | XP_006231337.1 | 10 | 1415 | |||
| XM_008760321.1 | XP_008758543.1 | 9 | 1311 | |||
| GenBank mRNA | BC072532.1 | - | 10 | 1448 | 7099460 | |
| S81497.1 | - | 10 | 1394 | |||

Lipa
Genomic Map