siRNA Details
| Gene Symbol: Avpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019136.1 | NP_062009.1 | 3 | 974 | ||
| XM_006229549.2 | XP_006229611.1 | 4 | 2368 | |||
| GenBank mRNA | AY338195.1 | - | 3 | 974 | ||
| Gene Symbol: Arhgap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY742900.1 | - | 3997 | |||
More Information
Set Membership: |
|

Avpr2
Genomic Map