siRNA Details
| Gene Symbol: Stat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034164.1 | NP_001029336.1 | 4, 5 | 554 | ||
| NM_032612.3 | NP_116001.2 | 4, 5 | 554 | |||
| GenBank mRNA | AF205604.1 | - | 2, 3 | 264 | ||
| BC062079.1 | - | 4, 5 | 554 | 5599645 | ||
| CV075250.1 | - | 4, 5 | 472 | |||

Stat1
Genomic Map