siRNA Details
| Gene Symbol: Egr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019137.1 | NP_062010.1 | 2 | 1538 | ||
| GenBank mRNA | M65008.1 | - | 2 | 1485 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Egr4| Gene Symbol: Egr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019137.1 | NP_062010.1 | 2 | 1538 | ||
| GenBank mRNA | M65008.1 | - | 2 | 1485 | ||