siRNA Details
| Gene Symbol: Igf2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012756.1 | NP_036888.1 | 40 | 5902 | ||
| GenBank mRNA | U59809.1 | - | 40 | 5902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igf2r| Gene Symbol: Igf2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012756.1 | NP_036888.1 | 40 | 5902 | ||
| GenBank mRNA | U59809.1 | - | 40 | 5902 | ||