siRNA Details
| Gene Symbol: Map1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030995.1 | NP_112257.1 | 4 | 7907 | ||
| XM_006234826.2 | XP_006234888.1 | 5 | 8277 | |||
| XM_006234827.2 | XP_006234889.1 | 5 | 8277 | |||
| XM_006234828.2 | XP_006234890.1 | 4 | 8181 | |||
| XM_008762145.1 | XP_008760367.1 | 4 | 7917 | |||
| GenBank mRNA | M83196.1 | - | 4 | 7907 | ||

Map1a
Genomic Map