siRNA Details
| Gene Symbol: Pgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022847.1 | NP_074038.1 | 9 | 2636 | ||
| GenBank mRNA | HM037361.1 | - | 3188 | |||
| HM037362.1 | - | 3188 | ||||
| L16922.1 | - | 9 | 2636 | |||
| S68283.1 | - | 3 | 243 | |||
| S68285.1 | - | 3 | 243 | |||

Pgr
Genomic Map