siRNA Details
| Gene Symbol: Cdkn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130812.3 | NP_570825.1 | 2 | 449 | ||
| XM_006238380.2 | XP_006238442.1 | 2 | 381 | |||
| XM_006238381.1 | XP_006238443.1 | 2 | 381 | |||
| GenBank mRNA | BC086995.1 | - | 2 | 384 | 7134283 | |
| CK603883.1 | - | 2 | 387 | |||
| S79760.1 | - | 2 | 424 | |||

Cdkn2b
Genomic Map