siRNA Details
| Gene Symbol: Cdkn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130812.3 | NP_570825.1 | 2 | 511 | ||
| XM_006238380.2 | XP_006238442.1 | 2 | 443 | |||
| XM_006238381.1 | XP_006238443.1 | 2 | 443 | |||
| GenBank mRNA | AF474978.1 | - | 2 | 425 | ||
| AF474979.1 | - | 2 | 427 | |||
| BC086995.1 | - | 2 | 446 | 7134283 | ||
| CK603883.1 | - | 2 | 449 | |||
| S79760.1 | - | 2 | 486 | |||

Cdkn2b
Genomic Map