siRNA Details
| Gene Symbol: Gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012767.2 | NP_036899.1 | 4 | 297 | ||
| XM_006252096.2 | XP_006252158.1 | 3 | 298 | |||
| GenBank mRNA | CR463456.1 | - | 1 | 172 | ||
| M12579.1 | - | 4 | 297 | |||
| S50870.1 | - | 3 | 271 | |||

Gnrh1
Genomic Map