siRNA Details
| Gene Symbol: Gnrh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012767.2 | NP_036899.1 | 2 | 56 | ||
| XM_006252096.2 | XP_006252158.1 | 1 | 57 | |||
| GenBank mRNA | M12579.1 | - | 2 | 56 | ||
| M15527.1 | - | 1 | 242 | |||
| S50870.1 | - | 1 | 30 | |||

Gnrh1
Genomic Map