siRNA Details
| Gene Symbol: Pcsk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017091.2 | NP_058787.1 | 1, 2 | 397 | ||
| GenBank mRNA | M76705.1 | - | 1, 2 | 399 | ||
| M83745.1 | - | 1, 2 | 341 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcsk1| Gene Symbol: Pcsk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017091.2 | NP_058787.1 | 1, 2 | 397 | ||
| GenBank mRNA | M76705.1 | - | 1, 2 | 399 | ||
| M83745.1 | - | 1, 2 | 341 | |||