siRNA Details
| Gene Symbol: Sry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012772.1 | NP_036904.1 | 1 | 255 | ||
| Gene Symbol: LOC103694545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773683.1 | XP_008771905.1 | 1 | 380 | ||
| Gene Symbol: LOC103694550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773684.1 | XP_008771906.1 | 1 | 416 | ||
| Gene Symbol: LOC103694552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773686.1 | XP_008771908.1 | 1 | 1337 | ||
| Gene Symbol: LOC103694554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773691.1 | XP_008771913.1 | 416 | |||
| Gene Symbol: LOC103694555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773692.1 | XP_008771914.1 | 1 | 416 | ||

Sry
Genomic Map