siRNA Details
| Gene Symbol: Aqp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012778.1 | NP_036910.1 | 1 | 75 | ||
| GenBank mRNA | BC090068.1 | - | 1 | 57 | 7190406 | |
| L07268.1 | - | 1 | 66 | |||
| X67948.1 | - | 1 | 75 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aqp1| Gene Symbol: Aqp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012778.1 | NP_036910.1 | 1 | 75 | ||
| GenBank mRNA | BC090068.1 | - | 1 | 57 | 7190406 | |
| L07268.1 | - | 1 | 66 | |||
| X67948.1 | - | 1 | 75 | |||