siRNA Details
| Gene Symbol: Nsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024128.3 | NP_077042.2 | 5 | 459 | ||
| GenBank mRNA | BC060538.1 | - | 5 | 462 | 6921928 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nsg1| Gene Symbol: Nsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024128.3 | NP_077042.2 | 5 | 459 | ||
| GenBank mRNA | BC060538.1 | - | 5 | 462 | 6921928 | |