siRNA Details
| Gene Symbol: Gamt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012793.2 | NP_036925.2 | 3, 4 | 439 | ||
| GenBank mRNA | FM043834.1 | - | 3, 4 | 412 | ||
| FQ210612.1 | - | 3, 4 | 362 | |||
| FQ225896.1 | - | 3, 4 | 439 | |||
| FQ227114.1 | - | 3, 4 | 439 | |||
| J03588.1 | - | 3, 4 | 437 | |||

Gamt
Genomic Map