siRNA Details
| Gene Symbol: Gstt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053293.2 | NP_445745.1 | 2 | 206 | ||
| XM_008772860.1 | XP_008771082.1 | 2 | 703 | |||
| GenBank mRNA | BC086426.1 | - | 2 | 206 | 7301492 | |
| FJ179408.1 | - | 2 | 176 | |||
| X67654.1 | - | 2 | 176 | |||

Gstt1
Genomic Map