siRNA Details
| Gene Symbol: Gstt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053293.2 | NP_445745.1 | 1 | 102 | ||
| GenBank mRNA | BC086426.1 | - | 1 | 102 | 7301492 | |
| FJ179408.1 | - | 1 | 72 | |||
| X67654.1 | - | 1 | 72 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gstt1| Gene Symbol: Gstt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053293.2 | NP_445745.1 | 1 | 102 | ||
| GenBank mRNA | BC086426.1 | - | 1 | 102 | 7301492 | |
| FJ179408.1 | - | 1 | 72 | |||
| X67654.1 | - | 1 | 72 | |||