siRNA Details
| Gene Symbol: Id1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012797.2 | NP_036929.2 | 1 | 282 | ||
| XM_006235267.2 | XP_006235329.1 | 1 | 1248 | |||
| GenBank mRNA | BC127448.1 | - | 1 | 294 | 8372348 | |
| D10862.1 | - | 1 | 263 | |||
| L23148.1 | - | 1 | 283 | |||
| M86708.1 | - | 1 | 289 | |||

Id1
Genomic Map