siRNA Details
| Gene Symbol: Pdgfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012802.1 | NP_036934.1 | 3 | 196 | ||
| XM_006250879.2 | XP_006250941.1 | 3 | 251 | |||
| XM_006250880.2 | XP_006250942.1 | 4 | 557 | |||
| XM_006250881.2 | XP_006250943.1 | 3 | 205 | |||
| GenBank mRNA | M63837.1 | - | 3 | 201 | ||

Pdgfra
Genomic Map