siRNA Details
| Gene Symbol: Pdgfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012802.1 | NP_036934.1 | 14 | 2083 | ||
| XM_006250879.2 | XP_006250941.1 | 14 | 2138 | |||
| XM_006250880.2 | XP_006250942.1 | 15 | 2444 | |||
| XM_006250881.2 | XP_006250943.1 | 14 | 2092 | |||
| GenBank mRNA | CK477487.1 | - | 1 | 34 | ||
| M63837.1 | - | 14 | 2088 | |||

Pdgfra
Genomic Map