siRNA Details
| Gene Symbol: Slc5a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230186.2 | XP_006230248.1 | 16 | 2536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a11| Gene Symbol: Slc5a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230186.2 | XP_006230248.1 | 16 | 2536 | ||