siRNA Details
| Gene Symbol: Rapgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761973.1 | XP_008760195.1 | 18 | 1800 | ||
| GenBank mRNA | AB031229.1 | - | 18 | 1976 | ||
| U78517.1 | - | 1 | 39 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rapgef4| Gene Symbol: Rapgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761973.1 | XP_008760195.1 | 18 | 1800 | ||
| GenBank mRNA | AB031229.1 | - | 18 | 1976 | ||
| U78517.1 | - | 1 | 39 | |||