siRNA Details
| Gene Symbol: Rapgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761973.1 | XP_008760195.1 | 28 | 2767 | ||
| GenBank mRNA | AB031229.1 | - | 28 | 2943 | ||
| U78517.1 | - | 11 | 1006 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rapgef4| Gene Symbol: Rapgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761973.1 | XP_008760195.1 | 28 | 2767 | ||
| GenBank mRNA | AB031229.1 | - | 28 | 2943 | ||
| U78517.1 | - | 11 | 1006 | |||