siRNA Details
Gene Symbol: Olr1413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214821.1 | NP_999986.1 | 1 | 643 | ||
GenBank mRNA | X89695.1 | - | 1 | 266 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214821.1 | NP_999986.1 | 1 | 643 | ||
GenBank mRNA | X89695.1 | - | 1 | 266 |