siRNA Details
| Gene Symbol: Sra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183329.3 | NP_899158.3 | 4, 5 | 501 | ||
| GenBank mRNA | AY026354.1 | - | 4, 5 | 449 | ||
| AY542868.1 | - | 4, 5 | 498 | |||
| BC091105.1 | - | 3, 4 | 1285 | 7381969 | ||
| CB322787.1 | - | 1, 2 | 441 | |||

Sra1
Genomic Map