siRNA Details
| Gene Symbol: Pcdhga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772071.1 | XP_008770293.1 | 1 | 1397 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhga4| Gene Symbol: Pcdhga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772071.1 | XP_008770293.1 | 1 | 1397 | ||