siRNA Details
| Gene Symbol: Tsga10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030022.1 | NP_001025193.1 | 4 | 268 | ||
| GenBank mRNA | AF092091.1 | - | 4 | 268 | ||
| BC168202.1 | - | 3 | 251 | 7115410 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsga10| Gene Symbol: Tsga10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030022.1 | NP_001025193.1 | 4 | 268 | ||
| GenBank mRNA | AF092091.1 | - | 4 | 268 | ||
| BC168202.1 | - | 3 | 251 | 7115410 | ||