siRNA Details
| Gene Symbol: Nr2c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145780.1 | NP_665723.1 | 2 | 221 | ||
| XM_008765216.1 | XP_008763438.1 | 2 | 868 | |||
| XM_008765217.1 | XP_008763439.1 | 3 | 585 | |||
| GenBank mRNA | BC061822.1 | - | 3 | 302 | 5599041 | |
| L26398.1 | - | 2 | 221 | |||

Nr2c1
Genomic Map