siRNA Details
| Gene Symbol: Cox5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145783.1 | NP_665726.1 | 1, 2 | 110 | ||
| GenBank mRNA | FQ210272.1 | - | 1, 2 | 143 | ||
| FQ211124.1 | - | 1, 2 | 170 | |||
| FQ217304.1 | - | 1, 2 | 143 | |||
| FQ217698.1 | - | 1, 2 | 133 | |||
| FQ223560.1 | - | 1, 2 | 143 | |||
| FQ223883.1 | - | 1, 2 | 127 | |||
| FQ224560.1 | - | 1, 2 | 135 | |||
| FQ224768.1 | - | 1, 2 | 136 | |||
| FQ228531.1 | - | 1, 2 | 143 | |||
| X15030.1 | - | 1, 2 | 110 | |||

Cox5a
Genomic Map