siRNA Details
| Gene Symbol: Cox5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145783.1 | NP_665726.1 | 3 | 284 | ||
| GenBank mRNA | FQ210272.1 | - | 3 | 317 | ||
| FQ211124.1 | - | 3 | 344 | |||
| FQ217304.1 | - | 3 | 317 | |||
| FQ217698.1 | - | 3 | 307 | |||
| FQ223883.1 | - | 3 | 301 | |||
| FQ224560.1 | - | 3 | 309 | |||
| FQ224768.1 | - | 3 | 310 | |||
| FQ228531.1 | - | 3 | 317 | |||
| X15030.1 | - | 3 | 284 | |||

Cox5a
Genomic Map