siRNA Details
Gene Symbol: Socs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145879.2 | NP_665886.2 | 1 | 151 | ||
GenBank mRNA | AW917497.1 | - | 2 | 152 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Socs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145879.2 | NP_665886.2 | 1 | 151 | ||
GenBank mRNA | AW917497.1 | - | 2 | 152 |