siRNA Details
| Gene Symbol: Cck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012829.2 | NP_036961.1 | 3 | 338 | ||
| XM_006244070.2 | XP_006244132.1 | 4 | 602 | |||
| GenBank mRNA | M25942.1 | - | 3 | 261 | ||
| X01032.1 | - | 3 | 338 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cck| Gene Symbol: Cck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012829.2 | NP_036961.1 | 3 | 338 | ||
| XM_006244070.2 | XP_006244132.1 | 4 | 602 | |||
| GenBank mRNA | M25942.1 | - | 3 | 261 | ||
| X01032.1 | - | 3 | 338 | |||