siRNA Details
| Gene Symbol: Cpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012836.2 | NP_036968.1 | 13 | 3072 | ||
| GenBank mRNA | AF284830.1 | - | 13 | 2429 | ||
| U62897.1 | - | 13 | 3072 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpd| Gene Symbol: Cpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012836.2 | NP_036968.1 | 13 | 3072 | ||
| GenBank mRNA | AF284830.1 | - | 13 | 2429 | ||
| U62897.1 | - | 13 | 3072 | |||