siRNA Details
| Gene Symbol: Cstb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012838.2 | NP_036970.1 | 2 | 213 | ||
| GenBank mRNA | BC084725.1 | - | 2 | 201 | 7318993 | |
| CF976177.1 | - | 2 | 214 | |||
| FQ220397.1 | - | 2 | 197 | |||
| FQ221069.1 | - | 2 | 173 | |||
| FQ222613.1 | - | 2 | 168 | |||
| X54737.1 | - | 2 | 205 | |||

Cstb
Genomic Map