siRNA Details
| Gene Symbol: Lhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033975.1 | NP_001029147.1 | 2 | 398 | ||
| NM_012858.2 | NP_036990.1 | 3 | 359 | |||
| XM_006228975.2 | XP_006229037.1 | 6 | 2164 | |||
| GenBank mRNA | BG379148.1 | - | 1 | 171 | ||
| CB566905.1 | - | 362 | ||||
| D00576.1 | - | 2 | 341 | |||
| U25653.1 | - | 6 | 2169 | |||
| U25802.1 | - | 6 | 2233 | |||
| V01542.1 | - | 3 | 351 | |||

Lhb
Genomic Map