siRNA Details
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012861.1 | NP_036993.1 | 5 | 546 | ||
GenBank mRNA | M76704.1 | - | 4 | 491 | ||
S61804.1 | - | 5 | 546 | |||
X54862.1 | - | 5 | 572 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012861.1 | NP_036993.1 | 5 | 546 | ||
GenBank mRNA | M76704.1 | - | 4 | 491 | ||
S61804.1 | - | 5 | 546 | |||
X54862.1 | - | 5 | 572 |