siRNA Details
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012862.1 | NP_036994.1 | 3 | 128 | ||
GenBank mRNA | J03026.1 | - | 3 | 128 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012862.1 | NP_036994.1 | 3 | 128 | ||
GenBank mRNA | J03026.1 | - | 3 | 128 |