siRNA Details
| Gene Symbol: Mgp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012862.1 | NP_036994.1 | 2, 3 | 101 | ||
| GenBank mRNA | BC086394.1 | - | 2, 3 | 133 | 7303359 | |
| FQ224403.1 | - | 2, 3 | 158 | |||
| J03026.1 | - | 2, 3 | 101 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgp| Gene Symbol: Mgp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012862.1 | NP_036994.1 | 2, 3 | 101 | ||
| GenBank mRNA | BC086394.1 | - | 2, 3 | 133 | 7303359 | |
| FQ224403.1 | - | 2, 3 | 158 | |||
| J03026.1 | - | 2, 3 | 101 | |||