siRNA Details
Gene Symbol: Bhlha15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012863.1 | NP_036995.1 | 2 | 494 | ||
XM_008768941.1 | XP_008767163.1 | 2 | 1155 | |||
GenBank mRNA | BC061868.1 | - | 2 | 442 | 5598799 | |
U58279.1 | - | 2 | 494 |
