siRNA Details
| Gene Symbol: Nfyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031553.2 | NP_113741.1 | 6, 7 | 733 | ||
| XM_006241196.2 | XP_006241258.1 | 6, 7 | 751 | |||
| GenBank mRNA | BC089791.1 | - | 6, 7 | 733 | 7380395 | |
| M55045.1 | - | 5, 6 | 602 | |||

Nfyb
Genomic Map