siRNA Details
| Gene Symbol: Thrsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012703.2 | NP_036835.2 | 1 | 130 | ||
| GenBank mRNA | BC089061.1 | - | 1 | 130 | 7308757 | |
| FQ214269.1 | - | 1 | 141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thrsp| Gene Symbol: Thrsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012703.2 | NP_036835.2 | 1 | 130 | ||
| GenBank mRNA | BC089061.1 | - | 1 | 130 | 7308757 | |
| FQ214269.1 | - | 1 | 141 | |||