siRNA Details
| Gene Symbol: Bcan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033665.1 | NP_001028837.1 | 5, 6 | 1017 | ||
| NM_012916.2 | NP_037048.2 | 5, 6 | 1017 | |||
| XM_006232595.2 | XP_006232657.1 | 5, 6 | 2098 | |||
| XM_006232596.2 | XP_006232658.1 | 5, 6 | 1001 | |||
| XM_006232597.2 | XP_006232659.1 | 5, 6 | 819 | |||
| GenBank mRNA | BC094307.1 | - | 1, 2 | 1797 | ||
| U37142.1 | - | 5, 6 | 821 | |||
| X79881.1 | - | 5, 6 | 871 | |||
| X86406.1 | - | 5, 6 | 871 | |||
| Z28366.1 | - | 5, 6 | 1012 | |||

Bcan
Genomic Map