siRNA Details
Gene Symbol: Dpysl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012933.1 | NP_037065.1 | 11, 12 | 1389 | ||
XM_008759973.1 | XP_008758195.1 | 8, 9 | 1000 | |||
GenBank mRNA | U52103.1 | - | 10, 11 | 1252 |
