siRNA Details
| Gene Symbol: Hbegf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012945.1 | NP_037077.1 | 1 | 52 | ||
| GenBank mRNA | L05489.1 | - | 1 | 52 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hbegf| Gene Symbol: Hbegf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012945.1 | NP_037077.1 | 1 | 52 | ||
| GenBank mRNA | L05489.1 | - | 1 | 52 | ||