siRNA Details
| Gene Symbol: Fabp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017098.1 | NP_058794.1 | 3, 4 | 374 | ||
| XM_008767638.1 | XP_008765860.1 | 2, 3 | 446 | |||
| GenBank mRNA | L22788.1 | - | 3, 4 | 374 | ||
| S52878.1 | - | 3, 4 | 353 | |||

Fabp6
Genomic Map